Staining Dish and Tray Set, Mortech®
Supplier: Mortech Manufacturing
BG002 BG003 BG004 BG005 BG006 BG007 BG008
76498-552EA 233.21 CAD
76498-552 76498-554 76498-556 76498-558 76498-560 76498-562 76498-564
Staining Dish and Tray Set, Mortech®
Staining Boxes
Histology slide staining tray and dish set is ideal for a wide variety of routine and special histology staining procedures.
  • Stainless steel construction
  • Capacity for 30 or 60 slides