Formalin Neutralizer Pads, Mortech®
Supplier: Mortech Manufacturing
BE093 BE094 BE095
76498-512EA 321.09 CAD
76498-512 76498-514 76498-516
Formalin Neutralizer Pads, Mortech®
Spill Control Materials
Dramatically minimizes exposure to formaldehyde vapors.
  • Minimizes cleanup
  • Incineration of pads is clean and efficient with minimal threat of exposure
  • Fiber absorbs and then locks in liquids so that work areas stay drier
  • Resists tearing, even when saturated
  • Internal sliding shelf for easy positioning cube for pouring and storage
  • Simple and easy to use
A major crisis facing today's hospitals, laboratories and other healthcare providers is the occupational exposure and disposal of formaldehyde, a known carcinogen. Formaldehyde Neutralizer Pads offer aMore Product Information