BxPads Blue Sponges, Mortech®
Supplier: Mortech Manufacturing
BI056
76498-608EA 78.64 CAD
76498-608
BxPads Blue Sponges, Mortech®
Wipes Sponges
BxPads are the thinnest biopsy sponges available.
Designed to minimize pressure applied to unfixed specimens in biopsy cassettes.
Packaging: 1000 pads per bag.