Coplin Staining Jar, Mortech®
Supplier: Mortech Manufacturing
BA002 BA003
76498-152EA 69.61 CAD
76498-152 76498-154
Coplin Staining Jar, Mortech®
Jars Staining Jars
Heavy, clear glass jar with four molded internal grooves to hold up to ten slides.
  • Glass base construction
  • Four molded grooves in glass hold up to ten 3×1" slides
Jar features a broad base to prevent spillage. Both glass and plastic lid options protect against contaminants. The plastic option is a tight-fitting screw lid to prevent fluid evaporation. The glass cover option simply and easily sets upon the jar.