StrepPRO™ Streptococcal Grouping Kit, Hardy Diagnostics
Supplier: Hardy Diagnostics
PL032HD PL033HD PL038HD PL040HD PL036HD PL031HD PL031HD PL037HD PL038HD PL033HD PL035HD PL034HD PL039HD PL037HD PL036HD PL030HD PL039HD PL032HD
CA10001-478EA 283.95 CAD
10324-482 10324-480 10324-380 10324-486 10324-484 75802-980 10324-488 10324-378 75802-982 CA10001-478 10063-762 10063-760 CA10001-486 CA10001-474 CA10001-476 CA10001-482 CA10001-484 CA10001-480
StrepPRO™ Streptococcal Grouping Kit, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
A unique streptococcal grouping latex agglutination kit for the rapid identification of group A, B, C, D, F, and G Streptococcus from culture.
  • Utilizes modified acid extraction technology
  • No incubation required
  • Greater than 98% sensitive with as little as one colony required
All color-coded reagents are ready-to-use and remain in one space-saving workstation.
Blue latex particles on a white reaction card with eight reaction circles create an exceptionally easy-to-read result.
Delivery information: Contains all reagents and reaction cards to perform 60 tests.