3B Scientific® Nikki the Nursing Manikin with Auscultation
Supplier: American 3B Scientific
1022952 1023318
470331-330EA 16097 CAD
470331-330 470359-168
3B Scientific® Nikki the Nursing Manikin with Auscultation
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Developed specifically for scenario-based simulation.
  • Select sound scenarios and change them remotely
  • Training of one-on-one student or patient interaction
  • User friendly software that enables maximum flexibility
Nikki the Nursing Manikin simulates an adult, life-size patient and comes with the NursingScope Auscultation Training Stethoscope powered by Cardionics. NursingScope is an electronic simulation-training stethoscope created for auscultation training purposes or objective structured clinical examinations (O.S.C.E.). It functions wirelessly with the included controller tablet and gives the instructor access to the large high quality sound library from Cardionics. The instructor can select and change any of the physiological and pathological sounds that the trainees will hear through the NursingScope once they place the stethoscope on one of the auscultation sites of the manikin.
The auscultation sounds library includes 21 heart sounds, 15 lungs sounds, and 6 bowel sounds.
The instructor can easily change the auscultation sounds on the controller tablet for each of the 15 locations using NursingScope’s wireless functionality. Thus, the instructor has a tool to adapt the scenario on the spot, so that it fits any nursing curriculum or training program. Nikki the Nursing Manikin can be set up both as a male or a female patient thanks to interchangeable genitals and can be positioned like a real patient with natural movement of the arms, legs, and joints. Nikki can even sit upright in the bed for treatments and examinations, making it the ideal training solution for bedside competencies. It is made of durable, unbreakable, and water-resistant plastics for longevity in the everyday training use.
Training with Nikki the Nursing Manikin PRO meets the OBRA requirements of Nurse Aide Training and Competency Evaluation Programs (NATCEP).
Please see below for a complete list of training possibilities:
Patient and personal care; Transfer skills: lifting, carrying, and moving a patient; Bed care of a patient; Position changing and alignment of a patient; Bathing a patient (genital care); Hair care (washing, combing, drying); Care and cleaning of eyes, ears and nose; Personal hygiene and mouth denture care (partially removable prosthesis); Bandaging (includes amputation stump); Wound treatment and care (surgical staple seam, laceration and abrasions); and Communication skills, especially step by step explanation of care measures Enema administration.
Medical care; prevention of bedsores (Decubitus); Decubitus evaluation (stages 2-4); decubitus care; prevention of muscle cramps; Prevention of pneumonia; prevention of blood clots (Thromboses); prevention of healthcare associated infection (HAI) by measures of medical asepsis and clean techniques; inhalation therapy; oxygen inhalation; use of ventilation bag (mouth to mouth and mouth to nose); artificial respiration, oxygen treatment; care of tracheostomy; care of diabetic foot syndrome (DFS); ostomy care (colostomy); irrigation of eyes and ears; intramuscular injections (buttocks); subcutaneous injections (upper arms and thighs); subcutaneous infusion.
The manikin also includes removable organs for additional patient care training features: Male catheterization CH14; Female catheterization CH 14; Gastric (stomach) lavage; intestinal irrigation; irrigation of the bladder; irrigation of the vagina.