Rapid Response™ Urine Adulteration Strip, BTNX

Supplier: BTNX
Rapid Response™
URA-1S65
76473-524KT 62.69 CAD
76473-524
Rapid Response™ Urine Adulteration Strip, BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Rapid Response® Urine adulteration test strip is a rapid, semi-quantitative, dip-and-read test for the determination of creatinine, nitrite, glutaraldehyde, pH, specific gravity, oxidants and Pyridium chlorochromate in urine specimens, to assess the integrity of a urine sample prior to drug screening.

  • Sample: Urine
  • Format: Strip
  • Time to result: Within 2 minutes
  • Storage condition: 15 to 30 °C (59 to 86 °F)
  • Test principle: Colorimetric analysis

Delivery information: Includes test strips, product insert.

Caution: This test is intended for in vitro diagnostic use only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR