Rapid Response™ Qualitative Urine Toxicology Control, BTNX
Supplier: BTNX
Rapid Response™
RR-1VPN-12.5
76473-502EA 268.52 CAD
76473-502
Rapid Response™ Qualitative Urine Toxicology Control, BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Rapid Response® Qualitative urine toxicology control is packaged in amber vials (borosilicate glass) with silicon septas. The matrix is synthetic urine.
  • Level 1 negative control solution (10 ml)
  • Level 2 positive control solution (10 ml)
This product lasts until the expiry date listed on the bottle, or 31 days after opening.
Delivery information: Includes 2 control solution vials (10 ml).
Caution: This test is for forensic use only.