Rapid Response™ hCG Pregnancy Test Cassette (Urine), BTNX

Supplier: BTNX INC.

HCG-1C25
76473-482KT 42.85 CAD
76473-482 76473-484
Rapid Response™ hCG Pregnancy Test Cassette (Urine), BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits

The Rapid Response® hCG Pregnancy test cassette is a rapid, qualitative test for the determination of human chorionic gonadotropin in human urine specimens, to aid in the early detection of pregnancy.


  • Sample: Urine
  • Format: Cassette
  • Specificity: Human Chorionic Gonadotropin
  • Accuracy: >99.9%
  • Time to result: 3 minutes
  • Storage condition: 2 to 30 °C (36 to 86 °F)
  • Test principle: Lateral flow immunoassay


Delivery information: Includes test devices, disposable pipettes, product insert.


Caution: This test is intended for in vitro diagnostic use only.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR