Rapid Response™ Urinalysis Reagent Strips, BTNX

Supplier: BTNX INC.

U10.1-1S100 U2.1-1S100 U2.3-1S25 U5.1-1S100 U7.1-1S100
76473-512KT 71.61 CAD
76473-512 76473-516 76473-518 76473-520 76473-522
Rapid Response™ Urinalysis Reagent Strips, BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits

The Rapid Response® Urinalysis reagent strip is a fast, qualitative and semi-quantitative, dip-and-read test for the detection of urine analytes in human urine, to aid in the general evaluation of health and in the diagnosis and monitoring of metabolic or systemic diseases.


  • Sample: Urine
  • Format: Strip
  • Time to result: Within 2 minutes
  • Storage condition: 2 to 30 °C (36 to 86 °F)
  • Test principle: Colorimetric analysis


Test results are interpreted by direct comparison to the color blocks printed on the canister label.


Urinalysis reagent strips can include combinations of leukocytes (LEU), nitrite (NIT), urobilinogen (URO), protein (PRO), pH, blood (BLO), specific gravity (SG), ketone (KET), bilirubin (BIL), glucose (GLU).


Delivery information: Includes reagent strips and product insert.


Caution: This test is intended for in vitro diagnostic use only.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR