NEBNext rRNA Depletion Kit v2 (Human, Mouse, Rat) with RNA Sample Purification Beads, New England Biolabs
Supplier: New England Biolabs (NEB)
E7405L E7405S E7405X
CA76447-134EA 1726 CAD
CA76447-134 CA76447-136 CA76447-138
NEBNext rRNA Depletion Kit v2 (Human, Mouse, Rat) with RNA Sample Purification Beads, New England Biolabs
Assays Nucleic Acid Assays
For added convenience, this kit contains NEBNext RNA Sample Purification Beads (Agencourt® RNAClean® XP beads from Beckman Coulter).
  • Superior depletion of rRNA from human, mouse and rat RNA
  • Depletion of cytoplasmic (5S, 5.8S, 18S, 28S, human ITS, ETS) and mitochondrial (12S and 16S) rRNA
  • Compatible with a broad range of input amounts: 10 ng - 1 µg
  • Suitable for low-quality or high-quality RNA
  • Fast workflow: 2 hours, with less than 10 minutes hands-on time
The NEBNext® rRNA Depletion Kit v2 features an improved probe design targeting all subunits of human pre-rRNA (5S, 5.8S, 18S, 28S, ITS, ETS), better coverage of mouse and rat mature rRNA, as well as improved enzyme formulations.