NEBNext rRNA Depletion Kit v2 (Human/Mouse/Rat), New England Biolabs
Supplier: New England Biolabs (NEB)
E7400L
E7400S
E7400X
CA76447-128EA
1657
CAD
CA76447-128
CA76447-130
CA76447-132
NEBNext rRNA Depletion Kit v2 (Human/Mouse/Rat), New England Biolabs
Assays
Nucleic Acid Assays
The NEBNext® rRNA Depletion Kit v2 features an improved probe design targeting all subunits of human pre-rRNA (5S, 5.8S, 18S, 28S, ITS, ETS), better coverage of mouse and rat mature rRNA, as well as improved enzyme formulations.
- Superior depletion of rRNA from human, mouse and rat RNA
- Depletion of cytoplasmic (5S, 5.8S, 18S, 28S, human ITS, ETS) and mitochondrial (12S and 16S) rRNA
- Compatible with a broad range of input amounts: 10 ng - 1 µg
- Suitable for low-quality or high-quality RNA
- Fast workflow: 2 hours, with less than 10 minutes hands-on time