OC-Light® S FIT Test Kits, Polymedco
Supplier: POLYMEDCO CANCER DIAGNOSTIC
FOB50S FOBSTRS FBPU-ABRMP120S FBPU-BULKS FBPUCUSTOM120S FBPU-NBRMPS FBPUS FOBBTLS FBPU-ADRS
75784-578KT 1501.92 CAD
75784-578 75784-750 75784-752 75784-754 75784-756 75784-758 75784-760 75784-762 75784-764
OC-Light® S FIT Test Kits, Polymedco
Clinical Diagnostic Supplies Clinical Reagents and Kits
OC-Light S FIT is a manual fecal immunochemical test for the detection of blood in stool that can lead to colorectal cancer.
  • Simple strip procedure with qualitative results in 5 minutes
  • Utilizes colloidal gold to enhance test line intensity and clarity
  • Requires no dietary or medicinal restrictions
Enables the physician or the patient to easily collect a sample and return the sample to the lab or physician office for quick and accurate results. Increased sensitivity, specificity, and patient compliance equates to less false positive results and more of the right patients being referred to colonoscopy.