Wright's stain
Supplier: AZER SCIENTIFIC CA
Synonyms:
Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
ES918
ES920
CA95057-508EA
1255.64
CAD
CA95057-508
CA95057-512
Wright's stain
Wright's stain
Wright stain is a polychromatic hematology stain consisting of eosin and methylene blue.
- Dye mixture produces multiple colors to clearly differentiate white blood cells, red blood cells appear red-orange
- Suitable for automated and manual staining protocols
- Wright stain can be used for urine cytology and fine needle aspirate cell preparations
MDL Number:
MFCD00082143 CAS Number: 68988-92-1 |