Eosin Y (yellowish) solution, ULTRA stain for histology
Supplier: AZER SCIENTIFIC CA
|
Danger
|
Synonyms:
Tetrabromofluorescein disodium salt
, Eosin Y disodium salt
, Eosin 225
, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt
, Solvent Red 43
, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate
, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)
, Field's Stain B
, 2,4,5,7-Tetrabromofluorescein disodium salt
Our Ultra Eosin-Y is specially formulated to provide a striking contrast and brilliant staining of all cytoplasmic components
- Provides familiar and vibrant cytoplasmic staining and differentiation
- It is alcoholic and acidified, a favorite of many pathologists and other end users
Azer Scientific Elite Histology Stains are formulated for consistently good performance on paraffin embedded or frozen section tissue.
For protocols requiring a clarifier, Elite Acid Rinse #1 is formulated to work with both the Prestige and Ultra stains.
Formula:
C₂₀H₆Br₄Na₂O₅ MW: 691.86 g/mol |
MDL Number:
MFCD00005040 CAS Number: 17372-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...