Hematoxylin, ULTRA stain for histology

Supplier: AZER SCIENTIFIC CA

Synonyms: (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaolHydroxybrasilinNSC 2785DcH-48Hydroxybrazilin(+)-HematoxylinNatural Black I

ES36101
CA71002-464CS 1414.34 CAD
CA71002-464
Hematoxylin, ULTRA stain for histology
Hematoxylin

Azer Scientific Elite Histology Stains are formulated for consistently good performance on paraffin embedded or frozen section tissue.


  • Hematoxylin does not require daily filtering
  • Lot to lot consistency
  • Premixed and ready to use
  • Suitable for cytology and histology labs


Elite Hematoxylin Stains offer consistent high quality nuclear chromatin deliniation with no affinity for background staining.


Ultra Stains deliver a traditional result familiar to many pathologists and end users. Ultra hematoxylin can be used in place of Gill stain for cytology applications.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR