Hematoxylin, Prestige stain for histology

Supplier: AZER SCIENTIFIC CA

Synonyms: (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaolHydroxybrasilinNSC 2785DcH-48Hydroxybrazilin(+)-HematoxylinNatural Black I

ES36001
CA71002-462CS 1632.5 CAD
CA71002-462
Hematoxylin, Prestige stain for histology
Hematoxylin

Azer Scientific Elite Histology Stains are formulated for consistently good performance on paraffin embedded or frozen section tissue.


  • Hematoxylin does not require daily filtering
  • Lot to lot consistency
  • Premixed and ready to use
  • Suitable for cytology and histology labs


Elite Hematoxylin Stains offer consistent high quality nuclear chromatin deliniation with no affinity for background staining.


The Prestige Hematoxylin stain solution produces results similar to Harris or Gill stain and Prestige Eosin-Y enhances cytoplasmic staining with three distinct color variations. Use Prestige Bluing to obtain the best cellular contrast.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR