Gram color staining kit stabilized
Supplier: AZER SCIENTIFIC CA
ES800
ES800
CA89133-283EA
537.14
CAD
89133-283
CA89133-283
Gram color staining kit stabilized
Gram color staining kit
Gram Stain Kit (stabilized Iodine) ready to use for the differentiation of bacteria
- Set consists of "pre-mixed" iodine
- For in vitro diagnostic use
- Specifically formulatess for differential staining of bacteria from cultures or patient specimens
Kit includes one 8 oz. bottle of crystal violet, stabilized iodine, decolorizer, and safranin.
A detailed product insert and convenient spout caps also included.