Gram's iodine solution stabilized

Supplier: BD CANADA MICROBIOLOGY CA

Synonyms: Potassium triiodideLugol's iodineIodine and potassium iodide

212542 212542
CA90000-710LPK 271.7 CAD
90000-710 CA90000-710L
Gram's iodine solution stabilized
Gram's iodine solution
Formula: I₃K
MW: 419.812 g/mol
MDL Number: MFCD00134128
CAS Number: 12298-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR