Acridine orange kit, BBL™ fluorescent dye
Supplier: BD CANADA MICROBIOLOGY CA
Synonyms:
Brilliant Acridine Orange E, 3,6-Bis(dimethylamino)acridine, Acridine Orange base, Solvent Orange 15, 3,6-Bis(dimethylamino)acridine N,N,N',N'-Tetramethyl-3,6-acridinediamine, Acridine orange free base, Acridine Orange Stain
BD BBL Acridine Orange Stain is recommended for use in the fluorescent microscopic detection of microorganisms in direct smears prepared from clinical and non-clinical materials. It is particularly useful in the rapid screening of normally sterile specimens, such as cerebrospinal fluid, where few organisms may be present and in the rapid examination of blood smears or smears containing proteinaceous material where differentiation of organisms from background material may be more difficult.
- Used for detecting microorganisms in direct smears by fluorescence.
Bacteria stain bright orange; human cells, tissue debris, and proteinaceous material stain pale green to yellow.
Formula:
C₁₇H₁₉N₃ MW: 265.35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005029 CAS Number: 494-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...