REDISHIP Intelli-Sense™ Multi-Speed Fiberglass Blowers, Labconco
Supplier: Lab Armor
718181210814 718181010814
76311-768EA 9804.69 CAD
76311-768 76311-766
REDISHIP Intelli-Sense™ Multi-Speed Fiberglass Blowers, Labconco
Ventilators
Intelli-Sense™ Multi-Speed Fiberglass Blowers are ideal for fume hood exhaust systems in moderate to highly corrosive conditions.
  • Three speeds easily adjusted by rocker switch
  • Withstands moderate to corrosive conditions
  • REDISHIP stock
They feature a direct drive ECM motor, which consumes one third less energy and is easier and less expensive to maintain than belt drive motors, and a phenolic-coated impeller and housing. Unit comes with control box and three speed blower switch. Adjustable over broad range of airflows.
The Intelli-Sense Blower provides an economical alternative to traditional on/off blowers. Users may close the sash and turn the Intelli-Sense Blower switch to standby mode so that the hood operates at a lower velocity yet fulfills minimum air exchange requirements. Lower velocity modes have the added benefit of quieter operation than full velocity mode.
They are not recommended for use with perchloric acid. Housings are made from molded fiberglass reinforced polyester for durability and long life. Balanced, forward-curved impellers made of injection-molded polypropylene resist corrosive atmospheres, operate quietly and provide optimum air delivery. Powder-coated base and weather cover provide long life in extreme weather.
Inlet and discharge diameters are sized to receive rigid PVC ducting material directly for easy installation. Thermal override protection automatically reduces RPM speeds when motor is overloaded. Flow-through vent holes and louvers allow air to circulate. Speed control box included for mounting on top of customer-supplied fume hood and blower switch and label for multiple speed operation. Variable exhaust orientation adjusts to eight outlet positions for installation flexibility (vertical discharge is recommended).
Certifications: UL listed motor.
Ordering information: Ductwork not included.