Giemsa's stain solution for hematology, used in parasitology

Supplier: AZER SCIENTIFIC CA

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

ES915-16 ES915
CA89130-204EA 485.27 CAD
CA89130-204 CA95057-502
Giemsa's stain solution for hematology, used in parasitology
Giemsa's stain

Giemsa stain is a classical blood film stain for peripheral blood smear and bone marrow specimens.


  • For the visualization of blood parasites


Erythrocytes stain pink, platelets show a light pale pink, lymphocyte and monocyte cytoplasm stain pale blue and leukocyte nuclear chromatin stains magenta.


Store at controlled room temperature (15-30°C). Keep bottle tightly closed when not in use. Do not use after expiration date specified on bottle label. Excessive precipitation in reagent is an indication of degradation

Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR