Thiazine solutions B stain for hematology (Wescor)
Supplier: AZER SCIENTIFIC CA
ES943
CA10746-660CS 379.06 CAD
CA10746-660
Thiazine solutions B stain for hematology (Wescor)
Thiazine solutions B
For use with for the Wescor® Aerospray stainer. Compatible with the ELITechGroup Aerospray® stainer
  • Ready to use
  • Lot to lot consistency
  • Biological Stain Commision certified dyes
Stain solutions B Thiazine (azure-B and methylene blue) is buffered to pH 6.8.