3B Scientific® CT Scanner
Supplier: American 3B Scientific
1017782 1017783 1017784 1017785
470233-746EA 12217.8 CAD
470233-746 470233-748 470233-750 470233-752
3B Scientific® CT Scanner
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Perform experiments in computed tomography.
  • Individual components allow you to purchase just what you need
  • CT sample designed for non-uniform Absorption
CT scanner for turning and moving samples for the purpose of generating tomographic images. All turning and movement is carried out by means of stepper motors. While the measurement is taking place, the scanner moves the sample backwards and forwards between the ultrasonic sensors connected to the outside of the sample container in accordance with the CT algorithm. Includes a sample stage for making recordings of suitable objects for observation. The sample stage is immersed in a sample trough. The whole slider is height adjustable so that it is possible for the area of the sample under investigation to be modified.