Hematoxylin solution Gill I stain for histology

Supplier: AZER SCIENTIFIC CA
Warning

Synonyms: HydroxybrasilinDcH-48Natural Black I

ES704-CA
CA11006-340CS 4701.76 CAD
CA11006-340
Hematoxylin solution Gill I stain for histology
Hematoxylin staining solutions

Gill #1 Hematoxylin progressive stain offers the flexibility for use in cytology applications or as a suitable counterstain in IHC protocols.


  • Dyes certified by the Biological Stain Commission
  • Small batch production
  • Lot to lot consitency


The single strength formulation optimally stains gynecological and non-gynecological specimens.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR