3B Scientific® Young's Modulus Apparatus
Supplier: American 3B Scientific
1018527 1018528
470233-674EA 2070.99 CAD
470233-674 470233-676
3B Scientific® Young's Modulus Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Measuring apparatus for investigating the elastic deformation of rods of flat geometry and for determining the modulus of elasticity.
  • Load characteristics of the strain gauge may be ignored
  • Measurements can be set up with the material samples supported on both sides or clamped at one end
Measuring apparatus for investigating the elastic deformation of rods of flat geometry and for determining the modulus of elasticity. Features a strain gauge unit for determining the deformation of electrically conducting material samples. The strain gauge unit is electrically connected to the material sample in such a way that placing of the probes on the sample is determined with a high degree of sensitivity and displayed with the help of two LEDs. The bending of the material sample with the weight suspended is measured to an accuracy of 0.01 mm and the modulus of elasticity can then be calculated from the reading.
Experiment Topics – Elastic deformation of flat bars; Determining modulus of elasticity.
Advantages – Load characteristics of the strain gauge may be ignored; Measurements can be set up with the material samples supported on both sides or clamped at one end.
Delivery information: Contents includes 6 Flat steel rods (W: 15 mm, L: 200 / 300 / 400 mm, thickness: 2 / 3 mm); 1 Strain gauge unit; 1 Horizontal beam with stand; 2 Knife-edge bearings; 1 Clamping chuck; 1 Set of weights and retaining clamps.