Alcohol 95%, Reagent Grade for histology (RGA 95)

Supplier: AZER SCIENTIFIC CA
Danger

Synonyms: Reagent alcohol

ES753 ES753
CA95041-466CS 1419.85 CAD
CA95041-466 95041-466
Alcohol 95%, Reagent Grade for histology (RGA 95)
Alcohol

Anhydrous reagent grade alcohol is a denatured form of ethyl alcohol, approved for sale under U.S. regulations (90 parts ethyl alcohol, 5 parts methyl alcohol, and 5 parts isopropyl alcohol).


Caution: This product can expose you to chemicals including Methanol, which is known to the State of California to cause birth defects or other reproductive harm

MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR