3B Scientific® Post Partum Hemorrhage Trainer
Supplier: American 3B Scientific
1021568 1021573 1021574 1021576 1021577 1021578
470311-934EA 2885.25 CAD
470311-934 470312-032 470312-034 470312-036 470312-038 470312-040
3B Scientific® Post Partum Hemorrhage Trainer
Models Human Biology Models
Train for complications during stage three and four labor.
  • Focuses on procedures immediately following birth
  • Simulate bleeding from three locations (vaginal, uterine, or cervical)
  • Easy to set up and adjust
  • The pump attachment allows easy adjustment of the uterine tone to simulate an atonic or contracted uterus
The PPH trainer is designed to be an affordable, versatile and realistic obstetric training tool, which will enhance the competence and the confidence of the healthcare professional providing postpartum care and managing postpartum bleeding.
The postpartum hemorrhage trainer offers accurate anatomy, including ischial spines, pubic bones and sacrum. For a realistic feel and texture in addition to a life-like softness, the vaginal canal and post-partum cervix have been made in SKINlike™ high-quality silicone.
The trainer will be useful for training the following procedures:
Delivery of placenta, also incomplete
Identification of the post-partum hemorrhage bleeding nature: uterus bleeding, cervix bleeding (rupture), vaginal bleeding (lesion)
Visual estimation of the blood loss volume up to 1,5l
Atonic uterus management
Practice the fundal massage
Practice bimanual compression
Practice catheterization with CH 14 catheter