3B Scientific® ORTHOBones
Supplier: American 3B Scientific
1019614 1019620 1017629
470311-740EA 47.2 CAD
470311-740 470311-742 470311-744 470311-746 470311-748 470311-750 470311-752 470311-754 470311-756 470311-758 470311-760 470311-762 470311-764 470311-766 470311-768 470311-770 470311-772 470311-774 470311-776 470311-778 470311-818 470311-820 470311-822 470311-842 470311-844 470311-846 470311-848 470311-850 470311-852 470311-884 470311-886 470311-888 470311-890 470311-892 470311-894 470311-896 470311-898 470311-900 470311-902 470311-904 470311-906 470311-950 470311-952 470311-954 470311-956 470311-958 470311-960 470311-962 470311-964 470311-966
3B Scientific® ORTHOBones
Models Human Biology Models
The next step in classroom osteology.
  • Simulated bone with a hard cortical shell and softer cancellous bone core
  • Perfect for students experiment
ORTHObones are a great tool for your students to use for experiments. The two layer construction gives each bone characteristics more like real bone while retaining a high level of structural detail. ORTHObones allow you to supply your students with a tool to practice medical engineering procedures, or a low cost model they can examine in greater detail.
Premium ORTHObones were developed for use by surgeons, orthopedists and medical engineers. These groups needed a bone with structural characteristics beyond what regular models could supply, but not at the cost of high anatomical details. ORTHObones meet all these needs. For a classroom, the cortical shell may be harder for students to perform procedures on. In this situation the Standard ORTHObones are a perfect fit. The standard grade was developed to give students in workshops or classrooms a product with the same two component structure, realistic haptics, anatomical accuracy, and a surface that is less rigid than the premium bones. This allows students to practice engineering implants, without the added difficulty of accurately breaking or cutting the heavier cortical bone.