Leybold Products Diode Pumped Nd: Yag Laser With Frequency Doubling
Supplier: KLINGER EDUCATION PROD CORP SE
P5863
470311-136EA 47497.99 CAD
470311-136
Leybold Products Diode Pumped Nd: Yag Laser With Frequency Doubling
Laser Pointers
Intra cavity iris for TEMoo operation.
  • Nonlinear optics
  • Phase matching condition
  • Clear demonstration of transverse models
  • Conversion efficiency
  • Dimensions: 48×36×24"
Properties of diode laser Nd: YAG crystal optical pumping rate equation model static and dynamic solution, laser output, power laser resonator, transversal modes cavity stability criterion, spiking Nd: YAG laser nonlinear optics phase matching condition, conversion efficiency, verification of quadratically relationship SHG power, clear demonstration of transverse modes intra cavity iris for TEMoo operation.
Principle of operation inside the cavity formed by the mirror (M1) and (M2) a KTP crystal is placed. The fundamental radiation is frequency doubled generating visible green radiation at the wavelength of 532 nm. The filter (F) blocks the pump as well as fundamental radiation in such a way that only green radiation can be characterized.