BD BBL™ MacConkey II Agar with Sorbitol, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
298519
CA-90005-144PK
305.53
CAD
CA-90005-144
CA90008-728
BD BBL™ MacConkey II Agar with Sorbitol, BD Diagnostics
Media for Microbiology
Prepared Media for Microbiology
The Prepared plated media MacConkey II agar with sorbitol effectively differentiates the serotype of Escherichia coli associated with hemorrhagic colitis (O157:H7) from most other nonpathogenic E. coli.