5(6)-TAMRA special formulation
Supplier: Anaspec Inc
Synonyms:
TAMRA 5 and 6-isomer, Tetramethylrhodamine-5-(and-6)-maleimide, 5(6)-Carboxy-tetramethylrhodamine, 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium inner salt
5(6)-TAMRA is the mixture of two carboxy tetramethylrhodamine (TMR) isomers. It is used to modify amino and hydroxy groups using EDC-mediated couplings when there are difficulties in using 5(6)-TAMRA, SE. TAMRA is one of the most popular fluorophores used in various bioconjugations.
- Spectral properties: Abs/Em = 541/565 nm
- Solvent system: DMF or DMSO
- Physical State: Solid
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: Blue ice
Formula:
C₂₅H₂₂N₂O₅ MW: 430.45 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16661194 CAS Number: 98181-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...