6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE)

Supplier: Anaspec Inc

AS-81008 AS-81008-100
103010-780EA 347.69 CAD
103010-780 103010-782
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE)
6-Carboxyfluorescein-N-succinimidyl ester (6-FAM SE)

6-FAM is another isomer of carboxyfluorescein. 6-FAM, SE is mainly used in sequencing of nucleic acids and labeling nucleotides.


  • Spectral properties: Abs/Em = 495/517 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Formula: C₂₅H₁₅NO₉
Storage Temperature: Freezer
MDL Number: MFCD00077323
CAS Number: 92557-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR