5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)
Supplier: Anaspec Inc
AS-81007
AS-81007-100
103010-774EA
284.62
CAD
103010-774
103010-776
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)
5-FAM, SE is a single isomer. It is one of the most popular green fluorescent reagents used for labeling peptides, proteins and nucleotides. It has also been used to prepare various small fluorescent molecules.
- Spectral properties: Abs/Em = 492/518 nm
- Solvent system: DMF or DMSO
- Appearance: Orange solid
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: Blue ice
Formula:
C₂₅H₁₅NO₉ Storage Temperature: Freezer |
MDL Number:
MFCD00077322 CAS Number: 92557-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...