5(6)-Carboxyfluorescein

Supplier: Anaspec Inc

Synonyms: 5(6)-FAM, 3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-ar-carboxylic acid

AS-81002 AS-81002-1
103010-746EA 181.54 CAD
103010-746 103010-748
5(6)-Carboxyfluorescein
5(6)-Carboxyfluorescein

Although FITC reagents have been more often used to prepare fluoresceinated bioconjugates, the low stability of FITC bioconjugates makes some researchers use amine-reactive succinimidyl esters of carboxyfluorescein (commonly called FAM) in bioconjugations. FAM reagents give carboxamides that are more resistant to hydrolysis. We have shown that FAM reagents require less stringent reaction conditions and give better conjugation yields, and the resulted conjugates have superior stability. We noted that FITC labeled peptides tend to deteriorate more quickly than the corresponding FAM conjugates.


  • Spectral properties: Abs/Em = 494/519 nm
  • Solvent system: DMF or DMSO
  • Appearance: Orange solid, Solvent System DMF or DMSO, used to prepare fluoresceinated bioconjugates
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: RT

Formula: C₄₂H₂₄O₁₄
MW: 752.63 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00151081
CAS Number: 72088-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR