7-Amino-4-methylcoumarin
Supplier: Anaspec Inc
AS-23482
AS-23483
103003-516EA
116.92
CAD
103003-516
103003-518
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin is used to produce fluorogenic substrates
- Spectral properties: Abs/Em = 351/430 nm
- Appearance: Solid
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: RT
Formula:
C₁₀H₉NO₂ Storage Temperature: Freezer |
MDL Number:
MFCD00006868 CAS Number: 26093-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...