4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)

Supplier: Anaspec Inc

AS-83210 AS-83211
103011-132EA 258.46 CAD
103011-132 103011-134
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)

AT-selective minor groove binder that exhibits nice fluorescence enhancement in the presence of DNA.


  • Spectral Properties Abs/Em = 358/461 nm
  • Solvent System: water
  • Storage: -20°C desiccated and protected from light

Formula: C₁₆H₁₅N₅·2HCl
Storage Temperature: Freezer
MDL Number: MFCD00012681
CAS Number: 28718-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR