Streptavidin
Supplier: Anaspec Inc
AS-60659
AS-60659-100
103005-956EA
372.31
CAD
103005-956
103005-958
Streptavidin
Proteins and Peptides
Streptavidin is a nonglycosylated, tetrameric protein, with each subunit able to bind a single molecule of the vitamin biotin. Streptavidin-biotin bond is the strongest known non-covalent interaction with Kd ~10-15 M. Because streptavidin lacks any carbohydrate modification and has a near-neutral pI, it has the advantage of much lower nonspecific binding than avidin. Streptavidin-biotin interaction is widely used in experimental biology for purification and detection of biomolecules.