CREBtide, FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
AS-60436-1
AS-60436-5
103003-136EA
641.54
CAD
103003-136
103003-138
CREBtide, FAM (Carboxyfluorescein)
Proteins and Peptides
CREBtide is a synthetic substrate for PKA (Km = 3.9 µM). This peptide is based on the phosphorylation sequence in δ-CREB (cAMP response element binding protein).
Sequence:5-FAM-KRREILSRRPSYR
MW:2075.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...