[Lys(Me1)36]-Histone H3 (21-44)-GK,Biotin

Supplier: Anaspec Inc

AS-64368-025 AS-64368-1
103008-012EA 309.23 CAD
103008-012 103008-014
[Lys(Me1)36]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides

This peptide is Histone H3 amino acid residues 21 to 44 mono-methylated at Lys-36 with an additional C-terminal glycine followed by a biotinylated lysine. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ATKAARKSAPATGGV-K(Me1)-KPHRYRPG-GK(Biotin)
MW:2931.5 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR