Histone H3 (21-44)-GK, Biotin
Supplier: Anaspec Inc
AS-64440-025
AS-64440-1
103008-158EA
310.77
CAD
103008-158
103008-160
Histone H3 (21-44)-GK, Biotin
Proteins and Peptides
This peptide is derived from Histone H3 21-44 amino acids, with additional glycine and lysine at the C-terminus with lysine biotinylated and used as substrate in methylation assays.
Sequence:ATKAARKSAPATGGVKKPHRYRPG-GK(Biotin)
MW:2917.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...