[Lys(Ac)9/14]-Histone H3 (1-21)-GGK,Biotin
Supplier: Anaspec Inc
AS-64363-025
AS-64363-1
103007-992EA
310.77
CAD
103007-992
103007-994
[Lys(Ac)9/14]-Histone H3 (1-21)-GGK,Biotin
Proteins and Peptides
This peptide is Histone H3 amino acid residues 1 to 21 acetylated at Lys-9 and Lys-14 with a C-terminal GG linker followed by a biotinylated lysine. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ARTKQTAR-K(Ac)-STGG-K(Ac)-APRKQLA-GGK(Biotin)
MW:2807.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...