[Lys(Me2)4]-Histone H3 (1-21)-GGK,Biotin

Supplier: Anaspec Inc

AS-64356-025 AS-64356-1
103006-532EA 309.23 CAD
103006-532 103008-086
[Lys(Me2)4]-Histone H3 (1-21)-GGK,Biotin
Proteins and Peptides

This peptide is Histone H3 amino acid residues 1 to 21 di-methylated at Lys-4 with a C-terminal GG linker followed by a biotinylated lysine. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ART-K(Me2)-QTARKSTGGKAPRKQLA-GGK(Biotin)
MW:2751.2 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR