Hyalophora cecropia Cecropin A Peptide
Supplier: Anaspec Inc
AS-24009
AS-24010
102999-782EA
455.38
CAD
102999-782
102999-784
Hyalophora cecropia Cecropin A Peptide
Proteins and Peptides
Cecropin A is a naturally occurring, linear, cationic, 37-residue antimicrobial peptide. Cecropin A kills bacteria by dissipating transmembrane electrochemical ion-gradients.
Sequence:KWKLFKKIEKVGQNIRDGIIKAGPAVAVVGQATQIAK-NH2
MW:4003.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...