Human Peptide YY (3-36)
Supplier: Anaspec Inc
AS-24405
AS-24406
102996-562EA
383.08
CAD
102996-562
102996-564
Human Peptide YY (3-36)
Proteins and Peptides
This peptide, a PYY (3-36), a Y2R agonist, is released from the gastrointestinal tract postprandially in proportion to the calorie content of a meal and can inhibit food intake.
Sequence:IKPEAPGEDASPEELNRYYASLRHYLNLVTRQRY-NH2
MW:4049.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...