Rat;Mouse Ghrelin Peptide

Supplier: Anaspec Inc

AS-24159 AS-24160
103003-682EA 641.54 CAD
103003-682 103003-684
Rat;Mouse Ghrelin Peptide
Proteins and Peptides

Rat/mouse Ghrelin differs from human Ghrelin on the 11th and 12th position - RV (Arg-Val) in rat/mouse and KA (Lys-Ala) in human. Ghrelin is an appetite stimulating peptide hormone secreted by stomach P/D1-type cells in humans and circulates in the bloodstream during fasting conditions.  Enzymatically n-octanoylated Serine3 of this 28-amino acid Ghrelin is the endogenous ligand specific for growth hormone secretagogue receptor 1a (GHSR1a). The GHSR1a receptor appears to be dedicated to Ghrelin and is widely expressed in different tissues. 
Sequence: GS-S(n-octanoyl)-FLSPEHQKAQQRKESKKPPAKLQPR
MW: 3314.8 Da
% Peak area by HPLC: 95
Storage condition: -20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR