Human Glucagon-Like Peptide 1, GLP-1 amide, FAM-labeled
Supplier: Anaspec Inc
This is fluorescent GLP-1 labeled at the peptide C-terminus with FAM, Abs/Em=494/521 nm. In response to Glucose ingestion, proglucagon in the intestinal L cells is cleaved into GLP-1 (1-36). Prior to secretion into the circulation, GLP-1 (1-36) is further processed into amidated GLP-1 (7-36)-and small amounts of glycine-extended GLP-1 (7-37). Both GLP-1 (7-36) and GLP-1 (7-37), causes glucose dependent release of insulin by pancreatic beta-cells. They also play a role in gastric motility (gastric emptying), on the suppression of plasma glucagon levels (glucose production) and possibly on the promotion of satiety and stimulation of glucose disposal in peripheral tissues independent of the actions of insulin. GLP-1 peptides such as GLP-1 (1-36) have been used to investigate restoration of pancreatic beta cell function. GLP-1 is also produced in the central nervous system.
Sequence: FAM-HDEFERHAEGTFTSDVSSYLEGQAAKEFIAWLVKGR-NH2
MW: 4469.8 Da
% Peak area by HPLC: 95
Storage condition: -20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...