Human Pancreatic Polypeptide, AnaSpec

Supplier: Anaspec Inc

AS-22866 AS-22867
102996-310EA 332.31 CAD
102996-310 102996-312
Human Pancreatic Polypeptide, AnaSpec
Proteins and Peptides

Pancreatic Polypeptide (PP) is a 36-amino acid anorexigenic peptide hormone secreted by the PP cells of pancreas as a response to hypoglycemia. It is rapidly released postprandially and continues to remain elevated for approximately 4-6 hours after a meal. PP modulates food intake, and is absent in children with Prader-Willi syndrome. Secretion of PP can be accomplished through electrical stimulation of the vagus nerve. Cholecystokinin (CCK) and Gastrin appear to be involved in its secretion, while Ghrelin, Obestatin and Somatostatin are reported to inhibit its release. PP is related to Peptide YY and Neuropeptide Y (NPY).
Sequence: APLEPVYPGDNATPEQMAQYAADLRRYINMLTRPRY-NH2
MW: 4181.8 Da
% Peak area by HPLC: 95
Storage condition: -20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR