Bovine Neurotensin
Supplier: Anaspec Inc
AS-22985
AS-24273
102996-356EA
413.85
CAD
102996-356
102996-492
Bovine Neurotensin
Proteins and Peptides
This 13 amino acid peptide was first isolated from bovine hypothalamus and was named from its neuronal localization. It was detected in the central nervous system (CNS) and peripheral tissues mainly in the gastrointestinal tract. This bioactive form of neurotensin post-translationally modified at a Glu residue was isolated from porcine intestine.
Sequence:Pyr-LYENKPRRPYIL
MW:1673 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...