Human;Pig;Rat Viasoactive intestinal peptide
Supplier: Anaspec Inc
AS-22872
AS-22873
102996-314EA
212.31
CAD
102996-314
102996-316
Human;Pig;Rat Viasoactive intestinal peptide
Proteins and Peptides
28-amino acid neuropeptide VIP (Vasoactive Intestinal Peptide) is a neurotransmitter and a neuromodulator.
Sequence:HSDAVFTDNYTRLRKQMAVKKYLNSILN-NH2
MW:3325.9 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...